Search
Search

TSPAN32| Gene Symbol: TSPAN32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021082612.1 | XP_020938271.1 | 708 | 50 | ||
| XM_021082613.1 | XP_020938272.1 | 710 | 50 | |||
| XM_021082614.1 | XP_020938273.1 | 113 | 50 | |||
Set Membership: |
|