Search
Search
| Gene Symbol: NLE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003358153.4 | XP_003358201.1 | 1023 | 60 | ||
| XM_005669035.3 | XP_005669092.1 | 878 | 60 | |||
| GenBank mRNA | AK234211.1 | - | 8 - 9 | 993 | 60 | |
Set Membership: |
|