Search
Search
| Gene Symbol: ACP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001928346.6 | XP_001928381.3 | 1123 | 95 | ||
| XM_021089952.1 | XP_020945611.1 | 1036 | 95 | |||
| GenBank mRNA | AK235757.1 | - | 5 - 6 | 1085 | 95 | |
| AK351385.1 | - | 5 - 6 | 1085 | 95 | ||
| AK396099.1 | - | 5 - 6 | 1107 | 95 | ||
| AY609695.1 | - | 5 - 6 | 955 | 95 | ||
Set Membership: |
|