Search
Search
| Gene Symbol: HLTF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003358605.5 | XP_003358653.2 | 604 | 86 | ||
| XM_013991989.2 | XP_013847443.1 | 604 | 86 | |||
| XM_013992040.2 | XP_013847494.1 | 610 | 86 | |||
| XM_013992063.2 | XP_013847517.1 | 605 | 86 | |||
| GenBank mRNA | AK236672.1 | - | 3 - 4 | 556 | 86 | |
Set Membership: |
|