Search
Search
| Gene Symbol: XRN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005669915.3 | XP_005669972.2 | 162 | 111 | ||
| GenBank mRNA | AK237874.1 | - | 1 - 2 | 162 | 111 | |
| AK348971.1 | - | 1 - 2 | 141 | 111 | ||
Set Membership: |
|