Search
Search
| Gene Symbol: ADCK5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005662812.3 | XP_005662869.1 | 220 | 79 | ||
| XM_005662813.3 | XP_005662870.1 | 184 | 79 | |||
| XM_013996404.1 | XP_013851858.1 | 296 | 79 | |||
| XM_021090463.1 | XP_020946122.1 | 220 | 79 | |||
| XM_021090464.1 | XP_020946123.1 | 182 | 79 | |||
| GenBank mRNA | AK343363.1 | - | 2 - 3 | 164 | 79 | |
Set Membership: |
|