Search
Search
| Gene Symbol: TAPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005666539.3 | XP_005666596.1 | 510 | 55 | ||
| XM_021101030.1 | XP_020956689.1 | 387 | 55 | |||
| XM_021101031.1 | XP_020956690.1 | 1391 | 55 | |||
| GenBank mRNA | AK346092.1 | - | 3 - 4 | 463 | 55 | |
Set Membership: |
|