Search
Search
| Gene Symbol: PHF10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005659097.3 | XP_005659154.2 | 890 | 61 | ||
| XM_021085631.1 | XP_020941290.1 | 896 | 61 | |||
| XM_021085636.1 | XP_020941295.1 | 987 | 61 | |||
| XM_021085638.1 | XP_020941297.1 | 981 | 61 | |||
| XM_021085643.1 | XP_020941302.1 | 1239 | 61 | |||
| XM_021085648.1 | XP_020941307.1 | 861 | 61 | |||
| XM_021085654.1 | XP_020941313.1 | 855 | 61 | |||
| GenBank mRNA | AK346362.1 | - | 7 - 8 | 857 | 61 | |
| AK347679.1 | - | 5 - 6 | 612 | 61 | ||
| AK396242.1 | - | 7 - 8 | 853 | 61 | ||
Set Membership: |
|