Search
Search
| Gene Symbol: MCOLN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003480808.4 | XP_003480856.1 | 545 | 70 | ||
| XM_021083918.1 | XP_020939577.1 | 552 | 70 | |||
| XM_021083919.1 | XP_020939578.1 | 550 | 70 | |||
| GenBank mRNA | AK346401.1 | - | 3 - 4 | 472 | 70 | |
| AK351251.1 | - | 3 - 4 | 493 | 70 | ||
Set Membership: |
|