Search
Search
| Gene Symbol: GPR180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003131046.4 | XP_003131094.1 | 842 | 157 | ||
| XM_005653818.3 | XP_005653875.1 | 643 | 157 | |||
| XM_021065783.1 | XP_020921442.1 | 842 | 157 | |||
| XM_021065784.1 | XP_020921443.1 | 842 | 157 | |||
| GenBank mRNA | AK348275.1 | - | 5 - 6 | 826 | 157 | |
Set Membership: |
|