Search
Search

KIAA0930| Gene Symbol: KIAA0930 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003481520.4 | XP_003481568.1 | 997 | 73 | ||
| XM_005663739.3 | XP_005663796.1 | 1129 | 73 | |||
| XM_013997469.2 | XP_013852923.1 | 1005 | 73 | |||
| XM_021091397.1 | XP_020947056.1 | 1092 | 73 | |||
| GenBank mRNA | AK348556.1 | - | 8 - 9 | 1161 | 73 | |
Set Membership: |
|