Search
Search
| Gene Symbol: RNGTT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021089928.1 | XP_020945587.1 | 902 | 84 | ||
| XM_021089934.1 | XP_020945593.1 | 889 | 84 | |||
| GenBank mRNA | AK349267.1 | - | 6 - 7 | 745 | 84 | |
Set Membership: |
|