Search
Search
| Gene Symbol: GRK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001924379.7 | XP_001924414.2 | 1166 | 81 | ||
| XM_013982990.2 | XP_013838444.1 | 944 | 81 | |||
| XM_021073437.1 | XP_020929096.1 | 996 | 81 | |||
| GenBank mRNA | AK232007.1 | - | 5 - 6 | 1065 | 81 | |
Set Membership: |
|