Search
Search
| Gene Symbol: PRKCD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013981774.2 | XP_013837228.1 | 906 | 72 | ||
| XM_021068957.1 | XP_020924616.1 | 1004 | 72 | |||
| XM_021068958.1 | XP_020924617.1 | 1018 | 72 | |||
| XM_021068959.1 | XP_020924618.1 | 900 | 72 | |||
| XM_021068960.1 | XP_020924619.1 | 792 | 72 | |||
| XM_021068961.1 | XP_020924620.1 | 1019 | 72 | |||
| XM_021068962.1 | XP_020924621.1 | 906 | 72 | |||
| XM_021068963.1 | XP_020924622.1 | 900 | 72 | |||
| XM_021068964.1 | XP_020924623.1 | 792 | 72 | |||
| XM_021068966.1 | XP_020924625.1 | 798 | 72 | |||
| GenBank mRNA | AK391562.1 | - | 3 - 4 | 257 | 72 | |
Set Membership: |
|