Search
Search
| Gene Symbol: CCDC47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021066771.1 | XP_020922430.1 | 744 | 132 | ||
| XM_021066772.1 | XP_020922431.1 | 733 | 132 | |||
| GenBank mRNA | AK393610.1 | - | 3 - 4 | 579 | 132 | |
Set Membership: |
|