Search
Search

METTL25| Gene Symbol: METTL25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021092800.1 | XP_020948459.1 | 564 | 85 | ||
| XM_021092802.1 | XP_020948461.1 | 564 | 85 | |||
| XM_021092803.1 | XP_020948462.1 | 564 | 85 | |||
| GenBank mRNA | AK395738.1 | - | 3 - 4 | 565 | 85 | |
Set Membership: |
|