Search
Search

CACNA1E| Gene Symbol: CACNA1E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021102614.1 | XP_020958273.1 | 5469 | 88 | ||
| XM_021102615.1 | XP_020958274.1 | 5518 | 88 | |||
| XM_021102616.1 | XP_020958275.1 | 5518 | 88 | |||
| XM_021102617.1 | XP_020958276.1 | 5506 | 88 | |||
| XM_021102618.1 | XP_020958277.1 | 5461 | 88 | |||
| XM_021102619.1 | XP_020958278.1 | 5428 | 88 | |||
| XM_021102620.1 | XP_020958279.1 | 5518 | 88 | |||
| XM_021102621.1 | XP_020958280.1 | 5461 | 88 | |||
| XM_021102622.1 | XP_020958281.1 | 5461 | 88 | |||
| XM_021102623.1 | XP_020958282.1 | 5518 | 88 | |||
Set Membership: |
|