Search
Search
| Gene Symbol: SYN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021091548.1 | XP_020947207.1 | 888 | 92 | ||
| XM_021091549.1 | XP_020947208.1 | 965 | 92 | |||
| XM_021091550.1 | XP_020947209.1 | 1009 | 92 | |||
| XM_021091552.1 | XP_020947211.1 | 991 | 92 | |||
Set Membership: |
|