Search
Search
| Gene Symbol: COG4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244337.1 | NP_001231266.1 | 12 - 13 | 1656 | 59 | |
| GenBank mRNA | AK235531.1 | - | 6 - 7 | 745 | 59 | |
| AK238244.1 | - | 1 - 2 | 113 | 59 | ||
| AK239873.1 | - | 1 - 2 | 39 | 59 | ||
| AK393772.1 | - | 12 - 13 | 1645 | 59 | ||
Set Membership: |
|