Search
Search

CYP4F55| Gene Symbol: CYP4F55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244636.1 | NP_001231565.1 | 2 - 3 | 325 | 89 | |
| XM_021079734.1 | XP_020935393.1 | 358 | 89 | |||
| XM_021079735.1 | XP_020935394.1 | 334 | 89 | |||
| XM_021079753.1 | XP_020935412.1 | 334 | 89 | |||
| GenBank mRNA | AK233210.1 | - | 2 - 3 | 89 | ||
| AK239242.1 | - | 2 - 3 | 325 | 89 | ||
| AK393150.1 | - | 2 - 3 | 283 | 89 | ||
Set Membership: |
|