Search
Search

cnn2| Gene Symbol: cnn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015796.1 | NP_001015796.1 | 6 - 7 | 692 | 73 | |
| XM_012953157.2 | XP_012808611.1 | 6 - 7 | 675 | 73 | ||
| GenBank mRNA | BC089746.1 | - | 6 - 7 | 692 | 73 | |
Set Membership: |
|