Search
Search

lysmd3| Gene Symbol: lysmd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017308.3 | NP_001017308.1 | 2 - 3 | 333 | 71 | |
| GenBank mRNA | BC158940.1 | - | 2 - 3 | 333 | 71 | |
| CR855525.2 | - | 361 | 71 | |||
Set Membership: |
|