Search
Search

aldh3a2| Gene Symbol: aldh3a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016537.2 | NP_001016537.1 | 6 - 7 | 981 | 94 | |
| XM_012956976.2 | XP_012812430.1 | 7 - 8 | 1111 | 94 | ||
| XM_012956977.2 | XP_012812431.1 | 7 - 8 | 1079 | 94 | ||
| XM_012956978.2 | XP_012812432.1 | 7 - 8 | 1110 | 94 | ||
| XM_012956979.2 | XP_012812433.1 | 7 - 8 | 1090 | 94 | ||
| XM_012956980.2 | XP_012812434.1 | 7 - 8 | 1061 | 94 | ||
| GenBank mRNA | BC091032.1 | - | 6 - 7 | 981 | 94 | |
| CR760779.2 | - | 6 - 7 | 981 | 94 | ||
Set Membership: |
|