Search
Search

tmprss4| Gene Symbol: tmprss4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045609.1 | NP_001039074.1 | 9 - 10 | 2181 | 64 | |
| XM_012966697.2 | XP_012822151.1 | 9 - 10 | 2250 | 64 | ||
| GenBank mRNA | CR855611.2 | - | 9 - 10 | 2181 | 64 | |
Set Membership: |
|