Search
Search

thap4| Gene Symbol: thap4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079353.1 | NP_001072821.1 | 5 - 6 | 1438 | 64 | |
| XM_012970575.2 | XP_012826029.1 | 4 - 5 | 630 | 64 | ||
| GenBank mRNA | BC121669.1 | - | 5 - 6 | 1438 | 64 | |
Set Membership: |
|