Search
Search

camk2a| Gene Symbol: camk2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100267.1 | NP_001093737.1 | 1 - 2 | 519 | 66 | |
| XM_012958394.1 | XP_012813848.1 | 1 - 2 | 662 | 66 | ||
| XM_012958396.1 | XP_012813850.1 | 1 - 2 | 662 | 66 | ||
| XM_012958398.2 | XP_012813852.1 | 1 - 2 | 665 | 66 | ||
| XM_012958400.1 | XP_012813854.1 | 1 - 2 | 662 | 66 | ||
| XM_012958401.1 | XP_012813855.1 | 1 - 2 | 666 | 66 | ||
| GenBank mRNA | BC135985.1 | - | 1 - 2 | 519 | 66 | |
Set Membership: |
|