Search
Search

Atg12| Gene Symbol: Atg12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144019.1 | NP_001137491.1 | 86 | 77 | ||
| GenBank mRNA | BT063792.1 | - | 353 | 77 | ||
| FJ445015.1 | - | 86 | 77 | |||
| FJ445016.1 | - | 86 | 77 | |||
Set Membership: |
|