Search
Search

ap17| Gene Symbol: ap17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001154904.1 | NP_001148376.1 | 520 | 128 | ||
| GenBank mRNA | BT060670.1 | - | 489 | 128 | ||
| EU959074.1 | - | 520 | 128 | |||
Set Membership: |
|