Search
Search

LOC100282445
LOC100383596| Gene Symbol: LOC100282445 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001155356.1 | NP_001148828.1 | 439 | 83 | ||
| GenBank mRNA | EU960930.1 | - | 439 | 83 | ||
| Gene Symbol: LOC100383596 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001176244.1 | NP_001169715.1 | 488 | 83 | ||
| GenBank mRNA | BT067415.1 | - | 416 | 83 | ||
| BT067731.1 | - | 488 | 83 | |||
Set Membership: |
|