Search
Search

pco106335| Gene Symbol: pco106335 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001152824.1 | NP_001146296.1 | 1180 | 68 | ||
| GenBank mRNA | BT055038.1 | - | 1195 | 68 | ||
| BT066275.1 | - | 1180 | 68 | |||
Set Membership: |
|