Search
Search

fie2| Gene Symbol: fie2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111712.1 | NP_001105182.1 | 1178 | 66 | ||
| GenBank mRNA | AY061965.1 | - | 1178 | 66 | ||
| AY103723.1 | - | 1178 | 66 | |||
| AY150643.1 | - | 105 | 66 | |||
| AY150644.1 | - | 984 | 66 | |||
| AY195849.1 | - | 788 | 66 | |||
| BT062314.1 | - | 1264 | 66 | |||
| DQ246046.1 | - | 1222 | 66 | |||
Set Membership: |
|