Search
Search

csu859(gol)| Gene Symbol: csu859(gol) | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001138361.1 | NP_001131833.1 | 813 | 71 | ||
| GenBank mRNA | BT033367.1 | - | 813 | 71 | ||
| BT035414.1 | - | 867 | 71 | |||
| EU963517.1 | - | 775 | 71 | |||
Set Membership: |
|