Bovine pulmonary artery endothelial cells. Alexa Fluor® 488 phalloidin, fluorescein and DAPI.
Photostability comparison of Alexa Fluor® 488 phalloidin (top panel, Cat. no. A12379) and fluorescein phalloidin (bottom panel, Cat. no. F432). Bovine pulmonary artery endothelial cells were labeled with 5 units/mL of Alexa Fluor® 488 phalloidin or fluorescein phalloidin, and 560 nM DAPI (blue-fluorescent nuclear staining, Cat. no. D1306, D3571, D21490). Images were acquired every three seconds for a total of 50 seconds using a Quantix camera (Photometrics).
Photostability comparison of Alexa Fluor® 488 phalloidin (top panel, Cat. no. A12379) and fluorescein phalloidin (bottom panel, Cat. no. F432).  Bovine pulmonary artery endothelial cells were labeled with 5 units/mL of Alexa Fluor® 488 phalloidin or fluorescein phalloidin, and 560 nM DAPI (blue-fluorescent nuclear staining, Cat. no. D1306, D3571, D21490).  Images were acquired every three seconds for a total of 50 seconds using a Quantix camera (Photometrics).
Related Products
Related Images