Insect Venom Allergen Facts, Symptoms, and Treatment
Learn more about common allergic diseases, symptoms, management paradigms, and testing considerations.
Is your patient a candidate for specific IgE testing?
Get detailed information on whole allergens and allergen components.
Ready to test a patient?
Learn more about common allergic diseases, symptoms, management paradigms, and testing considerations.
Is your patient a candidate for specific IgE testing?
Get detailed information on whole allergens and allergen components.
Ready to test a patient?
While many types of insects and insect-like bugs can cause allergic reactions, the most common varieties are bees, wasps, hornets, yellow jackets, and fire ants.1 Between 56.6 percent and 94.5 percent of the general population report being stung by a Hymenoptera insect (i.e., one that injects venom as a defense mechanism) at least once in their lifetimes.2 That said, it’s common for patients to be uncertain as to which type of insect inflicted the sting, which makes specific IgE blood testing particularly important.3
Typical sting reactions aren’t dangerous and include pain, inflammation, redness, and itching. However, more systemic reactions are possible and can be classified as mild, e.g., urticaria (hives), angioedema (swelling under the skin); moderate, e.g., gastrointestinal symptoms, dizziness, dyspnea (difficult or labored breathing); or severe, e.g., losing consciousness, respiratory or cardiac arrest, anaphylactic shock.2 In fact, up to an estimated 7 percent of the general population has experienced immediate systemic reactions to insect stings, and insect-venom allergy is among the most frequent triggers of severe anaphylaxis in adults.3,4
The most common insects that cause allergic reactions are bees, wasps, hornets, yellow jackets, and fire ants.1 Bumblebees are increasingly used for pollination in greenhouses, but allergy to their venom is still rare.2 While the ubiquitous honeybee usually leaves its stinger in its victim, where the stinger’s sac continuously pumps venom into the skin until it’s empty or removed, hornets, wasps, yellow jackets, and fire ants can sting repeatedly.3,4,5 Yellow jackets typically nest in the ground and in walls, but wasps and hornets tend to nest on buildings and in bushes and trees. Stinging insects are most active between late spring and early fall.5
Note that the same species of insects may have different names depending on their locales. For example, gallicus dominula, vulgaris germanica rufa, and media saxonica are all known as wasps in Europe, but in the United States, these species are referred to as paper wasps, yellow jackets, and hornets, respectively.3
Fire ants (aka imported fire ants) are native to South America, but they’re also found in Australia, North America, New Zealand, and several Caribbean, Asian, and European countries. The jack jumper ant and green-head ant are the predominant stinging varieties in Australia, where the former causes severe allergic reactions in 2 to 3 percent of sensitized individuals. In East Asia, a particular winged ant (i.e., Pachycondyla chinensis) poses a significant risk of anaphylaxis. While rare, severe reactions including anaphylaxis can also be caused by biting insects such as the Australian tick and kissing bug.7
The allergens of various honeybee species are similar, and bumblebee venom is comparable to that of honeybees. As such, both are highly cross-reactive.3 Additionally, certain properties of fire ant venom have some cross-reactivity with allergens from yellow jackets, honeybees, and wasps.7
Up to 59 percent of patients with venom allergies test positive for both honeybee and wasp venom. While some of these people have genuine double positivity (i.e., they’re sensitized to both types of venom), others may be experiencing cross-reactivity due to irrelevant cross-reactive carbohydrate determinants (CCD).8 In order to implement an effective management plan, it’s crucial to distinguish between double positivity and clinically irrelevant cross-reactivity.9
Knowing the proteins, or components, within each allergen that may be responsible for your symptoms can help distinguish what type of insect venom is the trigger. With that in mind, and based on your symptom history, your healthcare provider may suggest something called a specific IgE allergen component test to help diagnose your allergies. Results from this test can help your healthcare provider develop a management plan and decide if immunotherapy may reduce your symptoms.3
Your component test results will include the name of the components (a series of letters and numbers). Your healthcare provider will likely review the results with you, but here you’ll find an at-a-glance breakdown you can use as a reference. Simply match the component names to the list below to see what they mean in terms of symptom management.3,10,11
Api m 1, Api m 2, Api m 3, Api m 5, Api m 10 (honeybee)
Ves v 1, Ves v 5 (common wasp)
Pol d 5 (paper wasp)
MUXF3 (CCD)
Tryptase
*These products may not be approved for clinical use in your country. Please work with your healthcare provider to understand availability.
Your healthcare provider may recommend a plan that includes the following.2,5,12-14
Your healthcare provider may direct you to take one of the following medications or actions to improve your allergy symptoms:
Patients with venom allergy should carry an emergency kit including an adrenaline/epinephrine autoinjector.
If someone you’re with is having an allergic reaction and showing signs of shock, act quickly. Look for a weak, rapid pulse; trouble breathing; cool, clammy, and pale skin; confusion; and loss of consciousness. Immediately do the following:
Sting-reaction severity varies among individuals, but typical responses include pain, swelling, and redness confined to the sting site.5 Some people, however, will develop excessive local swelling, which can be immediate and peak within an hour or two after the sting or which can develop hours later, increase for two to 48 hours, and resolve after three to 10 days.4 Large local reactions where swelling extends beyond the site are also possible.5
Fire-ant stings can include an itchy localized lump at the sting site, which usually collapses within 30 to 60 minutes. The site then develops a small blister within four hours that eventually fills with dead tissue that resembles pus. However, the blister has a minimal chance of becoming infected unless it’s opened.5
A small percentage of the population has systemic or generalized reactions, which typically include the following mild, moderate, and severe reactions.2,4
Mild reactions usually affect the skin and subcutaneous tissues. They can include:4
Moderate reactions feature respiratory, gastrointestinal, or cardiovascular involvement and can include:4
Severe reactions can include:2,4
Insect-venom allergy can also cause anaphylaxis, which can include symptoms such as:6
Symptom severity can be amplified by various risk factors and co-factors, such as physical exertion, older age, presence of mast-cell disorders, and male gender.2 Of those adults with mild systemic reactions, roughly 10 percent have a more severe response with subsequent stings. That said, of those with moderate/severe reactions with the initial sting, approximately 45 percent have a less severe reaction with future stings.7 While rare, additional reactions may occur, including serum-sickness-like manifestations, Guillain-Barre syndrome, vasculitis, and more.2
Together with your symptom history, specific IgE blood testing can help determine if you are sensitized to a particular allergen. If you are diagnosed with an allergy, your healthcare provider will work with you to create a management plan.
*These products may not be approved for clinical use in your country. Please work with your healthcare provider to understand availability.
Venom allergy is one of the most frequent triggers of severe anaphylaxis in adults.3 An estimated 1 to 7 percent of the general population exhibits immediate systemic reactions to insect stings.4 Additionally, roughly 20 and 48 percent of severe anaphylaxis cases are caused by insect stings in children and adults, respectively. Anaphylactic death resulting from insect sting typically occurs within 15 to 20 minutes after exposure.2