    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • NMDAR2C Antibodies
          Antibodies Inc
          GluN2C/NR2C Monoclonal Antibody (N422/18), FL490
          View all (15) NMDAR2C antibodies
          Tech Support
          Cite GluN2C/NR2C Monoclonal Antibody (N422/18), FL490
          Additional Information:
          {{banner.description}}
          GluN2C/NR2C Monoclonal Antibody (N422/18), FL490
          Product Details
          75-411-FL490
          Applications
          Tested Dilution
          Publications
          Immunohistochemistry (Frozen) (IHC (F))
          Assay-dependent
          -
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          N422/18
          Immunogen
          Fusion protein amino acids 836-1233 (cytoplasmic C-terminus) of rat GluN2C (accession number Q00961) produced recombinantly in E. Coli.
          3D Epitope / Immunogen
          Conjugate
          FL490 FL490 FL490
          • Unconjugated
          • FL550
          • FL594
          • FL650
          • Request custom conjugation
          Excitation/Emission Max
          491/515 nm
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS
          Contains
          0.09% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Product Specific Information
          For maximum recovery of product, centrifuge the vial prior to removing the cap.
          Target Information
          The NMDA Receptor (NMDAR) plays an essential role in memory, neuronal development and it has also been implicated in several disorders of the central nervous system including Alzheimers, epilepsy and ischemic neuronal cell death. The NMDA receptor is also one of the principal molecular targets for alcohol in the CNS. The NMDAR is also potentiated by protein phosphorylation. The NR1 protein can form NMDA activated channels when expressed in Xenopus oocytes but the currents in such channels are much smaller than those seen in situ. Channels with more physiological characteristics are produced when the NR1 subunit is combined with one or more of th NMDAR2 (NR2 A-D) subunits.
          Bioinformatics
          Protein Aliases: GluN2C; GluN2C(alt_5'UTR_77nt); GluN2C(alt_5'UTR_87nt); GluN2C(del_e2); GluN2C-b alternative isoform; GluRepsilon3; Glutamate [NMDA] receptor subunit epsilon-3; glutamate receptor; Glutamate receptor ionotropic, NMDA 2C; glutamate receptor, ionotropic, N-methyl D-aspartate 2C; glutamate receptor, ionotropic, NMDA2C; N-methyl D-aspartate receptor subtype 2C; N-methyl-D-aspartate receptor subunit 2C; NMDA glutamate receptor; NMDAR2C; NR2; putative NMDtranscript(altAcc_e11); putative NMDtranscript(altDon_e4); putative NMDtranscript(del_e4)
          View more View less
          Gene Aliases: GluN2C; GRIN2C; NMDAR2C; NR2C
          View more View less
          UniProt ID: (Human) Q14957, (Mouse) Q01098, (Rat) Q00961
          View more View less
          Entrez Gene ID: (Human) 2905, (Mouse) 14813, (Rat) 24411
          View more View less
          Function(s)
          NMDA glutamate receptor activity ion channel activity cation channel activity protein binding ligand-gated ion channel activity signaling receptor activity transmitter-gated ion channel activity involved in regulation of postsynaptic membrane potential receptor activity ionotropic glutamate receptor activity extracellular-glutamate-gated ion channel activity PDZ domain binding protein N-terminus binding transmembrane signal receptor
          Process(es)
          ion transport glutamate receptor signaling pathway brain development response to wounding directional locomotion ion transmembrane transport ionotropic glutamate receptor signaling pathway synaptic transmission, glutamatergic negative regulation of protein catabolic process regulation of membrane potential regulation of synaptic plasticity regulation of neuronal synaptic plasticity neuromuscular process controlling balance positive regulation of synaptic transmission, glutamatergic excitatory postsynaptic potential long-term synaptic potentiation calcium ion transmembrane import into cytosol cation transmembrane transport excitatory chemical synaptic transmission protein localization to postsynaptic membrane regulation of cation transmembrane transport positive regulation of excitatory postsynaptic potential startle response transport protein localization
          Spain flag icon
          Spain
          TEST
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline