    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Synapsin 1/3 Antibodies
          Antibodies Inc
          Synapsin-1/3 Monoclonal Antibody (L125/129), FL650
          View all (5) Synapsin 1/3 antibodies
          Tech Support
          Cite Synapsin-1/3 Monoclonal Antibody (L125/129), FL650
          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:
          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          Synapsin-1/3 Monoclonal Antibody (L125/129), FL650
          Product Details
          75-497-FL650
          Applications
          Tested Dilution
          Publications
          Immunohistochemistry (Frozen) (IHC (F))
          Assay-dependent
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Mouse / IgG2b
          Class
          Monoclonal
          Type
          Antibody
          Clone
          L125/129
          Immunogen
          Fusion protein amino acids 76-417 (actin-binding and synaptic-vesicle binding domains) of human Synapsin-III (accession number O14994) produced recombinantly in E. Coli.
          3D Epitope / Immunogen
          Conjugate
          FL650 FL650 FL650
          • Unconjugated
          • FL490
          • FL550
          • FL594
          • Request custom conjugation
          Excitation/Emission Max
          655/676 nm
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS
          Contains
          0.09% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Product Specific Information
          For maximum recovery of product, centrifuge the vial prior to removing the cap.
          Target Information
          Synapsin 1 and Synapsin 3 are members of the synapsin family of proteins, which are involved in the regulation of neurotransmitter release at synapses. Both synapsin 1 and synapsin 3 are crucial for the proper functioning of synapses, playing key roles in neurotransmitter release, synaptic plasticity, and neuronal development.
          Bioinformatics
          Protein Aliases: ATP binding protein; similar to synapsin I and II; brain protein 4.1; C-terminal truncated due to the introduction of a pre-mature stop codon by the inclusion of intron 8; cN28H9.2 (synapsin III); neuronal phosphoprotein; SYN3; synapsin Ib; synapsin IIIa; synapsin-3-like; synaptic vesicle protein; unnamed protein product
          View more View less
          Gene Aliases: EPILX; EPILX1; MRX50; Syn-1; Syn1-S; SYN1a; SYN1b; SYNI
          View more View less
          UniProt ID: (Human) P17600, (Human) O14994, (Rat) P09951, (Mouse) O88935, (Rat) O70441, (Mouse) Q8JZP2
          View more View less
          Entrez Gene ID: (Human) 6853, (Human) 8224, (Rat) 24949, (Mouse) 20964, (Rat) 29130, (Mouse) 27204
          View more View less
          Function(s)
          protein binding ATP binding protein kinase binding identical protein binding microtubule cortical anchor activity calcium-dependent protein binding
          Process(es)
          chemical synaptic transmission neurotransmitter secretion regulation of neurotransmitter secretion modulation of synaptic transmission synapse organization synaptic vesicle clustering regulation of synaptic vesicle cycle synaptic vesicle cycle regulation of synaptic vesicle exocytosis neuron development regulation of short-term neuronal synaptic plasticity regulation of synaptic transmission, GABAergic
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Privacy Notice
          • Legal Notice
          Germany flag icon
          Germany
          Your items have has been added!
          Host server : magellan-search-blue-7fb9f846c9-wx6nt:80/100.66.72.150:80.
          git-commit: 8207b5afa5c305bd40e0a4f0b9b53fd46f2a8829
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.59.1-2026.09.91-1.0