    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • L-Myc Antibodies
          Proteintech
          MYCL1 Recombinant Rabbit Monoclonal Antibody (230104D6)
          View all (14) L-Myc antibodies
          Tech Support
          Cite MYCL1 Recombinant Rabbit Monoclonal Antibody (230104D6)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          MYCL1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          MYCL1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          MYCL1 Antibody (82881-1-RR) in ICC/IF
          Immunofluorescent analysis of (4% PFA) fixed U2OS cells using MYCL1 antibody (82881-1-RR) at dilution of 1:1,000 and Multi-rAb CoraLite ® Plus 594-Goat Anti-Rabbit Recombinant Secondary Antibody (H+L) (RGAR004). (CL488-Phalloidin, green). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          MYCL1 Antibody in Immunocytochemistry (ICC/IF)
          MYCL1 Antibody in Flow Cytometry (Flow)
          MYCL1 Recombinant Rabbit Monoclonal Antibody (230104D6)
          Product Details
          82881-1-RR
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:500-1:2,000
          -
          Flow Cytometry (Flow)
          0.25 µg/10^6 cells in a 100 µL suspension
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Expression System
          CHO cells
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          230104D6
          Immunogen
          MYCL1 fusion protein Ag30335
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • CoraLite Plus 488
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 50% glycerol
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: ERAVSDRLAP GAPRGNPPKA SAAPDCTPSL EAGNPAPAAP CPLGEPKTQA CSGSESPSDS
          Target Information
          Cellular DNA-binding proteins encoded by the c-myc genes. They are normally involved in nucleic acid metabolism and in mediating the cellular response to growth factors. Elevated and deregulated (constitutive) expression of c-myc proteins can cause tumorigenesis. L-Myc gene ontology annotations include DNA binding function, DNA-binding transcription factor activity, RNA polymerase II-specific action, protein dimerization activity, and regulation of inner ear auditory receptor cell differentiation.
          Bioinformatics
          Protein Aliases: bHLHe38; Class E basic helix-loop-helix protein 38; l-myc-1 proto-oncogene; myc-related gene from lung cancer; Protein L-Myc; Protein L-Myc-1; unnamed protein product; v-myc avian myelocytomatosis viral oncogene lung carcinoma derived homolog; V-myc myelocytomatosis viral oncogene homolog; v-myc myelocytomatosis viral oncogene homolog 1, lung carcinoma derived
          View more View less
          Gene Aliases: BHLHE38; L-Myc; LMYC; MYCL; MYCL1
          View more View less
          UniProt ID: (Human) P12524
          View more View less
          Entrez Gene ID: (Human) 4610
          View more View less
          Function(s)
          RNA polymerase II core promoter proximal region sequence-specific DNA binding RNA polymerase II transcription factor activity, sequence-specific DNA binding DNA binding transcription factor activity, sequence-specific DNA binding protein binding protein dimerization activity
          Process(es)
          regulation of transcription, DNA-templated regulation of transcription from RNA polymerase II promoter regulation of auditory receptor cell differentiation
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Privacy Information Center
          • Legal Notice
          Germany flag icon
          Germany
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline