Gas analysis applications for process mass spectrometers
Thermo Scientific process mass spectrometers for gas analysis are engineered to meet challenging applications in the biotechnology, chemical/petrochemical, clean energy, iron and steel, and semiconductor industries. Our process mass spec technologies help deliver faster, more complete, lab-quality online gas analysis and process analytics.
Bioprocessing / Biotech
Real-time gas analysis for optimized microbial and cell culture performance
Process mass spectrometers support advanced bioprocessing applications by providing continuous, high-precision gas analysis across microbial and mammalian cell processes. These insights are critical for maintaining consistency, optimizing yields, and ensuring process efficiency at every scale.
Thermo Scientific systems deliver real-time measurement of off-gases such as oxygen, carbon dioxide, nitrogen, and key volatiles. Scanning magnetic sector technology ensures fast, accurate analysis—helping track growth kinetics, substrate consumption, and process deviations without disrupting production.
With multi-stream capability, long calibration intervals, and seamless integration, our analyzers bring reliable, lab-quality gas analysis to both development and full-scale bioproduction environments.
Biopharma
Optimizing fermentation, cell culture, and API production through real-time gas analysis
Process mass spectrometers provide valuable insight into the physiological state of the culture. Analysis of respiratory gases is a non-invasive, real-time method for characterizing fermentation and cell culture processes. This approach reveals critical information—such as growth kinetics, substrate consumption, and overall culture health—and helps determine the optimal point to halt the process for maximum yield.
Our process mass spectrometers support pharmaceutical manufacturers in optimizing development, production, and quality control—from fermentation and mammalian cell cultures to solvent drying in active pharmaceutical ingredient (API) manufacturing.
Chemical/petrochemical
Optimizing production and maximizing profitability through real-time gas analysis
Fast, accurate, and comprehensive gas analysis enables real-time updates to control systems—helping optimize production units, reduce waste, and improve operational efficiency. Thermo Scientific process mass spectrometers meet the precision and accuracy standards required across the chemical and petrochemical industries.
Applications include ethylene furnace optimization, polyolefin production, monitoring fugitive VOC emissions, natural gas processing, ethylene oxide/ethylene glycol production, ammonia production, and more.
Flare stack emissions
Fast, multi-component analysis for accurate flare gas monitoring
Flare stacks burn off excess hydrocarbon gases that cannot be recovered or recycled, converting them into water vapor and carbon dioxide. With growing regulatory pressure, operators are now required to monitor the composition of flare gases in real time to minimize environmental impact and ensure compliance.
Thermo Scientific process mass spectrometers use scanning magnetic sector technology to deliver fast, flexible, and precise analysis of complex gas mixtures. Full flare gas composition is measured and reported within seconds, supporting efficient, compliant, and transparent operations.
This monitoring also enhances safety by detecting changes in flare gas that may signal process upsets. Ideal for oil refineries, chemical plants, and LNG facilities, these systems ensure operators stay ahead of emissions regulations while protecting both plant performance and the environment
Hydrogen
Real-time hydrogen measurement
During the production of green hydrogen by electrolysis, safety, process efficiency and product quality is of utmost importance, so the accurate analysis of hydrogen and other gases is critical for the successful creation of this potentially clean energy fuel. Our process mass spectrometers offer a robust range of features and are ideal tools for gas analysis in the production of green hydrogen.
Iron/steel
Real-time gas analysis to optimize every stage of steel production
Process mass spectrometers are essential in iron and steel manufacturing for their ability to deliver fast, accurate gas analysis across multiple process stages. From the blast furnace to the rolling mill, real-time data helps optimize combustion, improve efficiency, and maintain high product quality.
These systems analyze key gases such as CO, CO₂, N₂, and H₂ to optimize input materials and combustion in the blast furnace. Off-gas monitoring supports precise control of steel composition and impurity removal, while advanced gas cleaning oversight helps reduce pollutant emissions. In continuous casting and rolling mills, atmosphere control prevents oxidation and preserves steel quality. Emissions monitoring supports environmental reporting, and optimized fuel use leads to reduced energy consumption.
Thermo Scientific process mass spectrometers are built for the modern plant—offering broad component measurement on a single platform, advanced calibration, real-time data transmission, and self-diagnostic functionality for seamless integration and reliable performance.
Biofood
Gas analysis with mass spectrometry for scalable, sustainable food innovation
Biotechnology is reshaping food production, driving the development of enzyme-rich and protein-based alternatives through microbial fermentation. Enzymes once sourced from animal by-products or plants are now produced via biotech, enhancing food texture and flavor. Mycoprotein and other alternative proteins are also gaining ground as sustainable replacements for conventional meat, seafood, and dairy.
To scale these innovations, the biofood industry is adopting Process Analytical Technology (PAT)—a model first refined in pharmaceutical manufacturing. PAT centers on real-time monitoring of critical quality attributes (CQAs) and process parameters (CPPs) to optimize yield, efficiency, and quality.
Mass spectrometry plays a pivotal role in PAT for bio-based food production. Thermo Scientific process mass spectrometers deliver high-precision, continuous gas analysis of oxygen, carbon dioxide, nitrogen, argon, methanol, ethanol, and other volatiles. Integrated with a multi-stream sample selector, a single analyzer can monitor multiple fermentors, reducing implementation costs while accelerating production insight.
As the industry rises to meet both consumer demand and environmental responsibility, mass spectrometry empowers manufacturers to meet sustainability goals while maintaining uncompromising product integrity.
Semiconductors
Ultra-high sensitivity for uncompromising quality in semiconductor manufacturing
In advanced semiconductor production, even the smallest gas impurities can lead to performance failures, costly delays, and reduced yields. Thermo Scientific ultra-high purity gas analyzers deliver unmatched accuracy and lower detection limits than any other technology—empowering manufacturers to protect their processes in real time.
Designed for continuous online monitoring, these analyzers detect critical contaminants such as water and oxygen at single-digit parts per trillion (ppt) levels. This level of sensitivity ensures early detection and correction, safeguarding product integrity at every stage of manufacturing.
With fully automated operation and detection limits up to 100 times lower than traditional methods, our analyzers provide a powerful, cost-effective alternative to lab-based quality control—supporting the speed, precision, and reliability the semiconductor industry demands.
For Research Use Only. Not for use in diagnostic procedures.