Search
To acquire a suitable number of cells for research, a group of cells must first be disaggregated from a larger culture. Enter: cell dissociation. Gibco trypsin and alternative cell dissociation products are ideal for use with tissues and cell monolayers. Available in a variety of formats, both enzymatic and chemical, to meet the varied needs of researchers performing adherent cell culture. Explore the selection guide below to choose the correct cell dissociation reagent for your cell culture type.
| Cell culture type | Wash solution | Dissociation method | Recommended product | Format | Contains phenol red |
|---|---|---|---|---|---|
| Most continuous and strongly adherent cell lines | HBSS or PBS w/o calcium and magnesium | Trypsin 0.25% in balanced salt solution w/o calcium and magnesium | 0.25% Trypsin (1X) | Liquid | Yes |
| 2.5% Trypsin (10X) | Liquid | No | |||
| Trypsin (1:250) | Powder | No | |||
| Many cells at early passage | D-PBS | TrypLE reagent | TrypLE Express | Liquid | Yes |
| Continuous cell lines where cell surface protein integrity is important | HBSS or PBS w/o calcium and magnesium | Trypsin 0.05% in 0.53 mM EDTA | 0.05% Trypsin 0.53 mM EDTA | Liquid | Yes |
| 0.05% Trypsin 0.53 mM EDTA (10X) | Liquid | No | |||
| TrypLE reagent | TrypLE Express | Liquid | Yes | ||
| Weakly adherent epithelial cells | HBSS or PBS w/o calcium and magnesium | EDTA, glycerol in sodium citrate | Cell Dissociation Buffer, Hanks'-based | Liquid | No |
| Cell Dissociation Buffer, PBS-based | Liquid | No | |||
| Transformed fibroblasts | HBSS or PBS w/o calcium and magnesium | TrypLE reagent | TrypLE Express (1X) | Liquid | Yes |
| Primary cells where cell surface protein integrity is important | HBSS or PBS w/o calcium and magnesium | TrypLE reagent | TrypLE Express (1X) | Liquid | Yes |
| Strongly adherent early-passage cell lines | HBSS or PBS w/o calcium and magnesium | Trypsin 0.25% in 1 mM EDTA | 0.25% Trypsin 1 mM EDTA | Liquid | Yes |
| Trypsin Powder (1:250) | Powder | No | |||
| Dispase 0.6–2.4 units/mL in PBS | Dispase II | Powder, Lyophilized | No | ||
| Epithelial cells | 0.5–1 mM EDTA | 0.5–1 mM EDTA | Versene solution 1:5,000 (0.53 mM EDTA in PBS) | Liquid | No |
| Dispase 0.6–2.4 units/mL in PBS | Dispase II | Powder, Lyophilized | No | ||
| Strongly adherent, epithelial and some tumor cell lines | 0.5–1 mM EDTA | 0.25% with 1 mM EDTA | 0.25% Trypsin 1 mM EDTA | Liquid | Yes |
| Versene solution 1:5,000 (0.53 mM EDTA in PBS) | Liquid | No | |||
| Trypsin Powder (1:250) | Powder | No | |||
| Dispase 0.6–2.4 units/mL in PBS w/o calcium and magnesium | Dispase II | Powder, Lyophilized | No | ||
| Thick cultures, multiple layers | 1 mM EDTA | Trypsin 0.25% | 0.25% Trypsin 1 mM EDTA | Liquid | Yes |
| Trypsin Powder (1:250) | Powder | No | |||
| Dense cultures that are collagen rich | 1 mM EDTA | Collagenase 200 units/mL, 1 mM EDTA in balanced salt solution w/o calcium and magnesium | Collagenase Type I | Powder, Lyophilized | No |
| Collagenase Type II | Powder, Lyophilized | No | |||
| Collagenase Type IV | Powder, Lyophilized | No | |||
| All adherent cell cultures | HBSS or PBS w/o calcium and magnesium | Physical/mechanical | Cell scrapers | – | – |
Trypsin, a proteolytic enzyme, is the standard way to detach adherent cell cultures and monolayers. This globular, pancreatic protease cleaves at the C-terminal side of lysine and arginine, breaking down vessel-adhering proteins and allowing easy resuspension during cell harvesting.
Thermo Fisher Scientific offers porcine-derived trypsin, EDTA-enhanced trypsin, as well as formulations of the TrypLE enzyme, a gentle, trypsin-like enzyme. To avoid unplanned protein degradation, organic trypsin must be inactivated with trypsin inhibitors. In contrast, synthetic trypsin, such as Gibco TrypLE products, are deactivated via dilution, eliminating the need for additional solutions.
NOTE: If you have fetal bovine serum (FBS) in your cell culture media, the culture will need to be washed with a balanced salt solution before trypsinization to ensure trypsin functionality.
EDTA, a chelating agent, is commonly used in cell culture to improve trypsin’s ability to detach adherent cells. In cell dissociation solutions (like Trypsin-EDTA), EDTA binds calcium and magnesium into a hexadentate, weakening cell-to-cell adhesion and enhancing trypsin’s access to hydrolysis-targeted peptide bonds.
Gibco Trypsin-EDTA is made from irradiated porcine-derived protease with e-beam verification as well as tested for PPV, mycoplasma, and PCV 1/2 contamination.
Gibco TrypLE reagents are highly purified, recombinant cell-dissociation enzymes that replace porcine trypsin. These reagents are ideal for dissociating attachment-dependent cell lines in both serum-containing and serum-free conditions, and can be directly substituted for trypsin without protocol changes. TrypLE reagents are gentle on cells, stable at room temperature, and animal origin–free.
| Trypsin | Trypsin-EDTA | TrypLE Express | TrypLE Select | |
|---|---|---|---|---|
| Characteristics | Traditional standard | Trypsin with enhanced cleavage | Gentler, stabler, and cost comparable | Gentler, stabler, animal and human-component free |
| Use case | General purpose | General purpose | General purpose | Bioproduction/industrial application |
| Origin source | Porcine pancreas | Porcine pancreas | Fungus (animal origin-free) | Fungus (animal origin-free) |
| Manufacturing standards | cGMP manufacturing | cGMP manufacturing | cGMP manufacturing | cGMP, manufactured on dedicated animal origin-free machinery |
| Storage temperature | –5 to –20° C | –5 to –20° C | Room temperature | Room temperature |
| Higher cell viability | ✓ | ✓ | ||
| Inactivation method | Trypsin inhibitors | Trypsin inhibitors | Inhibition by dilution | Inhibition by dilution |
Trypsin inhibitors destructively alter trypsin, nullifying the proteolytic enzyme’s ability to bind with, and so digest, proteins. Used post dissociation, trypsin inhibitors protect your cells against further protein degradation by trypsin.
NOTE: Soybean trypsin inhibitors also bind chymotrypsin, albeit to a lesser degree.
| Soybean trypsin inhibitor (powder) | Defined trypsin inhibitor | Trypsin neutralizer solution | |
|---|---|---|---|
| Animal origin-free | ✓ | ||
| Calcium and magnesium free | ✓ | ||
| Sterile | ✓ | ✓ | |
| With 0.5% newborn bovine serum | ✓ |
Relatively gentle, collagenase works by digesting connective tissue’s native triple-helical collagen fibrils. These tissues are often found in skin, tendon, blood vessels, and bone—making collagenase disaggregation suitable for human tumor, mouse kidney, human brain, and epithelium cultures.
| Collagenase | Tissue/Cell type |
|---|---|
| Type I | Epithelial, adrenal, lung, fat |
| Type II | Heart, thyroid, salivary, liver, bone, cartilage |
| Type IV | Islet (insulin receptor sites) |
Dispase, or neutral protease, is a Bacillus polymyxa–produced metalloenzyme. Rapid and gentle, dispase can be used to harvest and transfer normal diploid cells and cell lines. Dispase effectively separates cell clumps and splits cells from intact tissue with little impact to membrane integrity or viability.
Versene solution, an EDTA solution in PBS, dissociates cells gently and non-enzymatically. This chelating agent detaches some cell types (such as epithelial cells) from vessel surfaces for suspension. It can also be used as a wash prior to trypsinization.
For further guidance on Versene solution applications, please see the cell dissociation selection guide by culture type.
Recommended for weakly adherent cells, such as epithelial cells, Gibco cell dissociation buffers dissociate gently and maintain surface protein integrity. Applications include cells involved in ligand binding, flow cytometry, and immunohistochemistry research.
Please consult the enzyme-free cell dissociation buffer protocol for more information.
We are dedicated to operational excellence—from how we manage our core quality processes to our risk management systems. Collaborate with Thermo Fisher Scientific for harmonized quality management, multi-site ISO certification, and robust controls designed to help safeguard your cell culture and cell therapy programs.
This detailed guide helps you find recycling programs and manage the end-of-use phase of your research products, including Gibco bottles. Where local recycling infrastructure exists, clean, uncontaminated Gibco bottles and caps may be recyclable through conventional plastic recycling streams aligned to their resin classification.
Keep your cells healthy while helping to reduce your lab’s environmental impact with greener cell culture products. Gibco, Thermo Scientific Nunc, and Invitrogen products can help advance sustainability in your lab by limiting the use of hazardous chemicals, reducing waste and material consumption, and increasing energy efficiency.
Cell Culture & Transfection Learning Center
Access cell culture and transfection educational resources for better experiment planning and execution.
Cell Culture Basics
Learn best practices and fundamentals of cell culture. Topics include aseptic technique and methods for the growth and maintenance of cells in culture.
Cell Culture Videos
Explore this collection of cell culture videos providing valuable insights into techniques, resources, and stay up to date with the latest advancements in the field.
Cell Culture Selection Tools
Find the right cell culture products for your research with these selection tools. Search for media, FBS, plastics, transfection reagents, and more.
Cell Culture & Transfection Support Center
Find technical support recommendations for your cell culture and transfection workflows, including tips for experimental setup and in-depth troubleshooting help.
Need technical support?
Contact our expert team for technical and application support of Laboratory Products
For Research Use Only. Not for use in diagnostic procedures.