ADVIA Centaur B·R·A·H·M·S PCT
ADVIA Centaur® B·R·A·H·M·S PCT may be used on these analyzers:
  • ADVIA Centaur® XP
  • ADVIA Centaur® CP 
The ADVIA Centaur® B·R·A·H·M·S PCT™ assay is a chemiluminescent  assay (CLIA) for the determination of procalcitonin (PCT) in human serum and plasma on the ADVIA Centaur® Immunoassay Systems  from Siemens Healthcare Diagnostics.
ADVIA Centaur B·R·A·H·M·S PCT is CE marked.
Assay Characteristics
 ADVIA Centaur B·R·A·H·M·S PCT assay can be distinguished by these characteristics:
Correlation (Centaur XP)
Correlation between Thermo Scientific™ B·R·A·H·M·S PCT™ sensitive KRYPTOR™ and ADVIA Centaur B·R·A·H·M·S PCT measured on ADVIA Centaur XP analyzer.
Correlation (Centaur CP)
Correlation between Thermo Scientific B·R·A·H·M·S PCT sensitive KRYPTOR and ADVIA Centaur B·R·A·H·M·S PCT measured on ADVIA Centaur CP analyzer.
Precision
Let us help unlock the potential of B·R·A·H·M·S PCT in your hospital.
Contact Us.
Thermo Fisher Scientific products are distributed worldwide; not all intended uses and applications are registered in every country. Availability of product in each country depends on local regulatory marketing authorization status.