Dimension EXL LOCI B·R·A·H·M·S PCT
The Dimension® EXL™ LOCI® B·R·A·H·M·S PCT™ assay is an in vitro diagnostic test for the quantitative measurement of procalcitonin (PCT) in human serum and plasma (lithium heparin, sodium heparin, K2EDTA, and K3EDTA) using the Siemens’ Dimension® EXL™ integrated chemistry system with LOCI® Module.
Dimension EXL LOCI B·R·A·H·M·S PCT is CE marked and FDA 510(k) cleared.
Assay Characteristics
Dimension EXL LOCI B·R·A·H·M·S PCT can be distinguished by these characteristics:
Limit of quantitation (LoQ) and limit of detection (LoD) values are rounded to two decimal places in alignment with the reported results for the assay.
Correlation 
Correlation between Dimension® EXL™ LOCI® B·R·A·H·M·S PCT and Thermo Scientific™ B·R·A·H·M·S PCT sensitive KRYPTOR™ (US IFU data)
Reproducibility* (US IFU data) 
a  Number of results.
b  Standard deviation.
c  Coefficient of variation.
* Reproducibility was determined in accordance with CLSI Document EP05‑A3
Let us help unlock the potential of B·R·A·H·M·S PCT in your hospital.
Contact Us.
Data available in Siemens Healthineers Dimension EXL LOCI BRAHMS Procalcitonin assay US IFU. Availability of products in each country depends on local regulatory marketing authorization status.