Lumipulse G B·R·A·H·M·S PCT
Lumipulse G B·R·A·H·M·S PCT may be used on these analyzers:
  • Lumipulse® G 600 II
  • Lumipulse® G 1200
Lumipulse® G B·R·A·H·M·S PCT is a chemiluminescent enzyme immunoassay (CLEIA) for the determination of procalcitonin (PCT) in human serum and plasma on the LUMIPULSE G systems from Fujirebio Inc.
LUMIPULSE G B·R·A·H·M·S PCT is CE marked.
Assay Characteristics
Lumipulse G B·R·A·H·M·S PCT assay can be distinguished by these characteristics:
Limit of quantitation (LoQ) and limit of detection (LoD) values are rounded to two decimal places in alignment with the reported results for the assay.
Correlation 
Correlation between Lumipulse G B·R·A·H·M·S PCT (Cartridge) and Thermo Scientific™ B·R·A·H·M·S PCT™ sensitive KRYPTOR™.
Precision
The Lumipulse G B·R·A·H·M·S PCT demonstrated precision ≤4% (total %CV) in a study run according to the Clinical and Laboratory Standards Institute (CLSI) Protocol EP5-A2.
Let us help unlock the potential of B·R·A·H·M·S PCT in your hospital.
Contact Us.
Thermo Fisher Scientific products are distributed worldwide; not all intended uses and applications are registered in every country. Availability of product in each country depends on local regulatory marketing authorization status.