LIAISON B·R·A·H·M·S PCT II GEN
LIAISON B·R·A·H·M·S PCT II GEN may be used on these analyzers:
  • LIAISON®
  • LIAISON® XL
  • LIAISON® XS
LIAISON® B·R·A·H·M·S PCT™ II GEN is a chemiluminescence immunoassay (CLIA) for the determination of procalcitonin (PCT) in human serum and plasma on the automated LIAISON random access analyzers from DiaSorin.
LIAISON B·R·A·H·M·S PCT II GEN is CE marked.
Assay Characteristics
LIAISON B·R·A·H·M·S PCT assay can be distinguished by these characteristics:
Limit of quantitation (LoQ) and limit of detection (LoD) values are rounded to two decimal places in alignment with the reported results for the assay.
Correlation 
Correlation between LIAISON B·R·A·H·M·S PCT II GEN and Thermo Scientific™ B·R·A·H·M·S PCT™ sensitive KRYPTOR™.
Reproducibility
The intra-assay and inter-assay coefficients of variation are <5% and <10%, respectively.
Let us help unlock the potential of B·R·A·H·M·S PCT in your hospital.
Contact Us.
Thermo Fisher Scientific products are distributed worldwide; not all intended uses and applications are registered in every country. Availability of product in each country depends on local regulatory marketing authorization status.