ELECSYS B·R·A·H·M·S PCT
ELECSYS B·R·A·H·M·S PCT may be used on these analyzers:
  • Roche Elecsys 8000 Modular Series
  • Roche Elecsys Cobas e411
  • Roche Elecsys Cobas e601
  • Roche Elecsys Cobas e602
  • Roche Elecsys Cobas e801 
Elecsys B·R·A·H·M·S PCT is an electrochemiluminescence immunoassay (ECLIA) for the determination of procalcitonin (PCT) in human serum or plasma on the automated Roche Elecsys®, Modular and Cobas e (e411, e601, e602 and e801) immunoassay analyzers.
Elecsys® B·R·A·H·M·S PCT is CE marked and FDA 510(k) cleared.
Assay Characteristics
ELECSYS B·R·A·H·M·S PCT assay can be distinguished by these characteristics:
Functional sensitivity and limit of detection (LoD) values are rounded to 2 decimal places in alignment with the reported results for the assay.
Correlation 
Correlation between ELECSYS B·R·A·H·M·S PCT and Thermo Scientific™ B·R·A·H·M·S PCT™ sensitive KRYPTOR™. 
Precision
Reproducibility was determined using Elecsys reagents, pooled human serum/plasma, and controls in accordance with the protocol EP5-A2 of the CLSI (Clinical and Laboratory Standards Institute): 2 runs per day in duplicates each for 21 days (n=84).
Elecsys 2010 and Cobas e411 analyzers
Modular Analytics E170 and Cobas e601 analyzers
Cobas e801 analyzer
Let us help unlock the potential of B·R·A·H·M·S PCT in your hospital.
Contact Us.
Thermo Fisher Scientific products are distributed worldwide; not all intended uses and applications are registered in every country. Availability of product in each country depends on local regulatory marketing authorization status.