B·R·A·H·M·S PCT sensitive KRYPTOR may be used on these analyzers:
The Thermo Scientific™ B·R·A·H·M·S PCT ™ sensitive KRYPTOR™ is a homogeneous immunoassay for the quantitative determination of PCT (Procalcitonin) in human serum or plasma on the automated Thermo Scientific B·R·A·H·M·S KRYPTOR random access analyzer.
B·R·A·H·M·S PCT sensitive KRYPTOR is CE marked and FDA 510(k) cleared.
Assay Characteristics
The B·R·A·H·M·S PCT sensitive KRYPTOR assay can be distinguished by these characteristics:
Correlation
Correlation - B·R·A·H·M·S PCT sensitive KRYPTOR vs. B·R·A·H·M·S PCT™ LIA™.
Reproducibility
*Inter-assay CV: Between calibrations, same calibrator and kit lot, same machine."
Thermo Fisher Scientific products are distributed worldwide; not all intended uses and applications are registered in every country.
Availability of product in each country depends on local regulatory marketing authorization status.