Search
Search
Antibodies Inc
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
For maximum recovery of product, centrifuge the vial prior to removing the cap.
Sequence of this protein is as follows: EIDSILGNKP YSKKDYRS
KVbeta1, KVbeta2, and KVbeta3 are beta subunits of voltage-gated potassium (KV) channels. These auxiliary subunits modulate the properties and functions of the KV channel's alpha subunits. These KVbeta subunits are essential for the fine-tuning of KV channel activity, impacting the electrical properties of cells and contributing to the regulation of physiological processes such as neuronal signaling and cardiac rhythm.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alternative splice product of the Shaker beta 1 gene, see GenBank Accession Numbers U16953 and L39833; hKvbeta2; K(+) channel subunit beta-1; K(+) channel subunit beta-2; K(+) channel subunit beta-3; K+ channel Beta1a chain; KCNAB2; Kv-beta-1; Kv-beta-2; Kv-beta-3; Neuroimmune protein F5; potassium channel beta 3 chain; potassium channel beta3 subunit; potassium channel shaker chain beta 1a; potassium channel subunit; potassium channel, voltage gated shaker-related subfamily A regulatory beta subunit 2; potassium channel, voltage gated shaker-related subfamily A regulatory beta subunit 3; potassium channel, voltage gated subfamily A regulatory beta subunit 1; potassium channel, voltage gated subfamily A regulatory beta subunit 2; potassium channel, voltage gated subfamily A regulatory beta subunit 3; potassium channel, voltage-dependent, beta-3 subunit; potassium voltage gated channel shaker related subfamily beta member 1; potassium voltage gated channel, shaker related subfamily, beta member 1; potassium voltage gated channel, shaker related subfamily, beta member 2; potassium voltage gated channel, shaker related subfamily, beta member 3; potassium voltage-gated channel subfamily A member regulatory beta subunit 1; potassium voltage-gated channel, shaker-related subfamily, beta member 1; potassium voltage-gated channel, shaker-related subfamily, beta member 2; potassium voltage-gated channel, shaker-related subfamily, beta member 3; RCK beta3; shaker-related subfamily member 1; unnamed protein product; voltage-gated potassium channel beta subunit; Voltage-gated potassium channel subunit beta-1; Voltage-gated potassium channel subunit beta-2; Voltage-gated potassium channel subunit beta-3
Gene Aliases: AKR6A3; AKR6A5; AKR6A9; Akr8a8; C330022D06Rik; Ckbeta2; Ckbeta3; F5; hKvb3; HKvbeta2; HKvbeta2.1; HKvbeta2.2; hKvBeta3; I2rf5; KCNA1B; KCNA2B; KCNA3.1B; KCNA3B; KCNAB1; KCNAB2; KCNAB3; Kcnab4; Kcnb3; KCNK2; KV-BETA-1; KV-BETA-2; KV-BETA-3; Kvb1; Kvb1.3; Kvbeta1.1; KvBeta3; mKv(beta)1; mKv(beta)4
UniProt ID: (Human) Q14722, (Human) Q13303, (Human) O43448, (Mouse) P63143, (Rat) P63144, (Mouse) P62482, (Rat) P62483, (Rat) Q63494
Entrez Gene ID: (Human) 7881, (Human) 8514, (Human) 9196, (Mouse) 16497, (Rat) 29737, (Mouse) 16498, (Rat) 29738, (Rat) 58981, (Mouse) 16499
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support