Please consider the latest version of this product introducing the most recent chemistry and protocol improvements for best performance. For more information, visit A57283 product page
Ion AmpliSeq™ HD Library Kit
Ion Torrent™

Ion AmpliSeq™ HD Library Kit

The Ion AmpliSeq HD Library Kit is designed for generation of amplicon libraries for use as part of the IonRead more
Have Questions?
Catalog NumberQuantity
A3769424 Reactions
Catalog number A37694
Price (USD)
4,600.00
Each
Quantity:
24 Reactions
Price (USD)
4,600.00
Each

The Ion AmpliSeq HD Library Kit is designed for generation of amplicon libraries for use as part of the Ion AmpliSeq HD sequencing workflow. The kit is compatible with custom panels created using the Ion AmpliSeq Designer tool (www.ampliseq.com) and sequenced using an Ion GeneStudio S5 Series instrument. Ion AmpliSeq HD technology enables scalable polymerase chain reactions (PCRs) from 12- to 5000-plex (1 x 5000-plex in a 1-pool panel or 2 x 2500-plex in a 2-pool panel) for Unique Molecular Tag (UMT)-targeted sequencing, with sample input amounts as small as 1 ng and a level of detection down to 0.1% depending on the sample source.

Key product features include:

  • Custom design creation
  • Ultra-high sensitivity detection
  • Dual-barcoded sample multiplexing
  • Bi-directional sequencing

Custom design creation

The Ion AmpliSeq Designer tool can now be used to create custom designs for use with Ion AmpliSeq HD technology. The designs are made-to-order and allow the user the flexibility of creating region designs, hotspot designs, or fusion designs. These designs are one- or two-pool designs and recommended for use with samples obtained from cfDNA , cfRNA, cfTNA, or FFPE.

Ultra-high sensitivity detection

Ion AmpliSeq HD technology is capable of ultra-high levels of detection down to 0.1%. These detection levels have been observed for cfDNA controls with known variants using an input amount of 20 ng. Levels of detection for other sample types, such as FFPE, will vary.

Dual-barcoded sample multiplexing

The Ion AmpliSeq HD Library Kit includes reagents for generating amplicons using Ion AmpliSeq HD primers, which are used to prepare libraries for sequencing. The libraries are dual-barcoded at the 5' and 3' ends using the Ion AmpliSeq HD Dual Barcode Kit 1-24, which enables the multiplexing of up to 24 samples in a single chip.

Bi-directional sequencing

The Ion AmpliSeq HD Library Kit also provides sequencing information from both strands. This enables better error correction and higher confidence in the quality of your sequence data.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Sequencing
For Use With (Equipment)Ion GeneStudio™ S5 Plus System, Ion GeneStudio™ S5 Prime System, Ion GeneStudio™ S5 System
LibrariesTargeted Sequencing Library
No. of Reactions24
Product LineAmpliSeq
Product TypeHD Library Kit
Quantity24 Reactions
Sample TypeDNA
Shipping ConditionDry Ice
Workflow StepLibrary Generation
FormatKit
Sequencing TypeGenome and DNA Sequencing
Unit SizeEach
Contents & Storage
1 x Amplification Mix (4X)
1 x SUPA Reagent
1 x CRC
2 x Low TE

Store at -5 to -30°C.

Frequently asked questions (FAQs)

How long can 10X DNA working panel FWD and REV subpools be stored?

10X DNA working panel FWD and REV subpools can be stored at 4 degrees C for one week. For longer term storage, aliquot and store subpools at -20 degrees C.

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.

Have there been any recent changes to the protocol for Ion AmpliSeq HD Library Kit with HD Enhancer (Cat. No. A57283)?

There have been some recent changes to the protocol. Please refer to the latest user guide (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0017392_Ion%20AmpliSeqHDLibraryKit_UG.pdf) for full details.

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.

When should I use CRC versus HD Enhancer for Ion AmpliSeq HD library preparation?

CRC is a component of the Ion AmpliSeq HD Library Kit (Cat. No. A37694). CRC improves target amplification reactions for challenging panels that produce excessive amounts of primer dimer or panels that have ⋝500 primer pairs. We recommend using CRC with all panel types. HD Enhancer is a component of the Ion AmpliSeq HD Library Kit with HD Enhancer (Cat. No. A57283) and is also available separately as Cat. No. A53690. HD Enhancer is used for improved library quality and molecular coverage results. Guidelines for using HD Enhancer or CRC for Ion AmpliSeq HD library preparation are outlined in the user guide (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0017392_Ion%20AmpliSeqHDLibraryKit_UG.pdf).

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.

Which barcodes are compatible with Ion AmpliSeq HD Library Kit with HD Enhancer (Cat. No. A57283)?

We recommend using the Ion AmpliSeq HD Dual Barcode Kit 1-24 (Cat. No. A37695) with Ion AmpliSeq HD Library Kit with HD Enhancer (Cat. No. A57283). The libraries are dual barcoded at the 5' and 3' ends and this enables the multiplexing of up to 24 samples in a single chip. Using these dual barcodes also reduces the risk of barcode cross-contamination.

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.

What are the shipping and storage conditions for Ion AmpliSeq HD Library Kit with HD Enhancer (Cat. No. A57283)?

Ion AmpliSeq HD Library Kit with HD Enhancer (Cat. No. A57283) is shipped on dry ice and we recommend storing it at -20 degrees C.

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.