Search
Search

| Assay Name: | bta-miR-584 |
|---|---|
| miRBase Accession Number: | MI0009874 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGGUUUGCCUGGGACUGAG |
| Species: | Bovine |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000533, mir-584 |
Mature miRNA Sequence: | UGGUUUGCCUGGGACUGAG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Bovine | bta-miR-584 | MIMAT0009352 |