Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MH15345
          Assay Name: bta-miR-584
          miRBase Accession Number: MI0009874
          miRBase Version: v22.1
          Chromosome Location: -
          Mature miRNA Sequence: UGGUUUGCCUGGGACUGAG
          Species: Bovine
          Product Type: mirVana® miRNA inhibitor
          Assay ID MH15345

          Purification
          Size
          Availability Made To Order
          Catalog # 4464084
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000533, mir-584

          Mature miRNA Sequence:

          UGGUUUGCCUGGGACUGAG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Bovine bta-miR-584 MIMAT0009352

          Stem-loop Details



          Bovine

          Stem-loop ID bta-mir-584-1
          Stem-loop Accession # MI0009874
          Stem-loop Sequence
          UCGGGUGAUCAACCGUCAUGGUUUGCCUGGGACUGAGGGGUUUCUUGGAAUGCAAGGCUUCCAGUGCUCAAGCCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009352
          miRBase ID: bta-miR-584
          Mature miRNA Sequence: UGGUUUGCCUGGGACUGAG
          Chromosome Location: NA

          Stem-loop ID bta-mir-584-2
          Stem-loop Accession # MI0009868
          Stem-loop Sequence
          UAGGGUGUCCAAGCCCUGCUGGUUUGCCUGGGACUGAGGAGUUUCUCAGGCUAAAGCCAGGAAAGUUCAGGGCAAACCAGGGUGAGCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009352
          miRBase ID: bta-miR-584
          Mature miRNA Sequence: UGGUUUGCCUGGGACUGAG
          Chromosome Location: NA

          Stem-loop ID bta-mir-584-3
          Stem-loop Accession # MI0009869
          Stem-loop Sequence
          UAGGGUGACCAACUAUCCUGGUUUGCCUGGGACUGAGAAAUUUCCUGGGAUGCAGAGCUUUCAGUACUAUAUAGCCUGGACAGUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009352
          miRBase ID: bta-miR-584
          Mature miRNA Sequence: UGGUUUGCCUGGGACUGAG
          Chromosome Location: NA

          Stem-loop ID bta-mir-584-4
          Stem-loop Accession # MI0009870
          Stem-loop Sequence
          GACCAUCCUCCUGGUUUGCCUGGGACUGAGUGGUUUCCUGGGUCUGUGGGACUUCCAGUGCUUAAACUGGGCAGGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009352
          miRBase ID: bta-miR-584
          Mature miRNA Sequence: UGGUUUGCCUGGGACUGAG
          Chromosome Location: NA

          Stem-loop ID bta-mir-584-5
          Stem-loop Accession # MI0009871
          Stem-loop Sequence
          AGGGAUGACCAACUGUCCUGGUUUGCCUGGGACUGAGGGGUUACUGAGGACACGUGAAGUCCCUGGGCAAACUGGAGUGGUUGGUUCAUUCUAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009352
          miRBase ID: bta-miR-584
          Mature miRNA Sequence: UGGUUUGCCUGGGACUGAG
          Chromosome Location: NA

          Stem-loop ID bta-mir-584-6
          Stem-loop Accession # MI0009872
          Stem-loop Sequence
          UUGGAUGACCAAUCAUCUUGGUUUGCCUGGGACUGAGGGGUUUCCCUGGAUGUGGGAUGUGUUGAAGCCAGGCAUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009352
          miRBase ID: bta-miR-584
          Mature miRNA Sequence: UGGUUUGCCUGGGACUGAG
          Chromosome Location: NA

          Stem-loop ID bta-mir-584-7
          Stem-loop Accession # MI0009873
          Stem-loop Sequence
          AGGGUGACCAACUGUCUUGGUUUGCCUGGGACUGAGGGUUUCCUGGGAUGUGGGAUGUUUAGGACUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009352
          miRBase ID: bta-miR-584
          Mature miRNA Sequence: UGGUUUGCCUGGGACUGAG
          Chromosome Location: NA

          Stem-loop ID bta-mir-584-8
          Stem-loop Accession # MI0010453
          Stem-loop Sequence
          AGGAUGGCCAAGCAUUCUGGUUUGCCUGGGACUGAGUGCCUGAGGUACGGGACUUGCAGUGCCAAGAUUAGGAUGUUGUUGUUGCUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009352
          miRBase ID: bta-miR-584
          Mature miRNA Sequence: UGGUUUGCCUGGGACUGAG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ MicroRNA Assay : 006045_mat
          mirVana® miRNA mimic : MC15345

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0