Search
Search

| Assay Name: | hsa-miR-153-3p |
|---|---|
| Stem-loop Accession Number: | MI0000463 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UUGCAUAGUCACAAAAGUGAUC |
| Species: | Human, Mouse, Rat, Alligator mississippiensis, Astatotilapia burtoni, Bovine, Callithrix jacchus, Capitella teleta, Columba livia, Fugu rubripes, Goat, Guinea pig, Horse, Lottia gigantea, Melibe leonina, Metriaclima zebra, Monodelphis domestica, Neolamprologus brichardi, Oreochromis niloticus, Ornithorhynchus anatinus, Pan troglodytes, Pongo pygmaeus, Pundamilia nyererei, Salmo salar, Taeniopygia guttata, Tetraodon nigroviridis, Zebrafish |
| Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000050, mir-153 |
Mature miRNA Sequence: | UUGCAUAGUCACAAAAGUGAUC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-153-3p | MIMAT0000439 |
| Mouse | mmu-miR-153-3p | MIMAT0000163 |
| Rat | rno-miR-153-3p | MIMAT0000855 |
| Alligator mississippiensis | ami-miR-153-3p | MIMAT0038184 |
| Astatotilapia burtoni | abu-miR-153a | MIMAT0042081 |
| Bovine | bta-miR-153 | MIMAT0009239 |
| View More Rows | ||
| Callithrix jacchus | cja-miR-153 | MIMAT0039527 |
| Capitella teleta | cte-miR-153 | MIMAT0009528 |
| Columba livia | cli-miR-153a-3p | MIMAT0038546 |
| Fugu rubripes | fru-miR-153a | MIMAT0003011 |
| Goat | chi-miR-153 | MIMAT0035983 |
| Guinea pig | cpo-miR-153-3p | MIMAT0047035 |
| Horse | eca-miR-153 | MIMAT0012936 |
| Lottia gigantea | lgi-miR-153 | MIMAT0009582 |
| Melibe leonina | mle-miR-153-3p | MIMAT0048756 |
| Metriaclima zebra | mze-miR-153a | MIMAT0042315 |
| Monodelphis domestica | mdo-miR-153-3p | MIMAT0012738 |
| Neolamprologus brichardi | nbr-miR-153a | MIMAT0042492 |
| Oreochromis niloticus | oni-miR-153a | MIMAT0042692 |
| Ornithorhynchus anatinus | oan-miR-153-3p | MIMAT0007005 |
| Pan troglodytes | ptr-miR-153 | MIMAT0008047 |
| Pongo pygmaeus | ppy-miR-153 | MIMAT0002272 |
| Pundamilia nyererei | pny-miR-153a | MIMAT0042892 |
| Salmo salar | ssa-miR-153b-3p | MIMAT0032391 |
| Taeniopygia guttata | tgu-miR-153-3p | MIMAT0014552 |
| Tetraodon nigroviridis | tni-miR-153a | MIMAT0003012 |
| Zebrafish | dre-miR-153a-3p | MIMAT0001849 |